Assay Details
Gene Symbol: LIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001245923.1 | NP_001232852.1 | 205 | 99 | ||
XM_017350650.1 | XP_017206139.1 | 231 | 99 | |||
GenBank mRNA | AX797954.1 | - | 205 | 99 | ||
AX798851.1 | - | 205 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
