Assay Details
Gene Symbol: CD40LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256781.1 | NP_001243710.1 | 4 - 5 | 412 | 97 | |
GenBank mRNA | JN635525.1 | - | 4 - 5 | 412 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
