Assay Details
Gene Symbol: AKR1B10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267588.1 | NP_001254517.1 | 7 - 8 | 798 | 70 | |
GenBank mRNA | AB724112.1 | - | 7 - 8 | 798 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
