Assay Details
Gene Symbol: MYC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319575.1 | NP_001306504.1 | 1 - 2 | 754 | 82 | |
XM_008255902.2 | XP_008254124.1 | 2 - 3 | 999 | 82 | ||
XM_017340986.1 | XP_017196475.1 | 2 - 3 | 1144 | 82 | ||
GenBank mRNA | KJ708483.1 | - | 1 - 2 | 754 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
