Assay Details
Gene Symbol: TNFSF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001329086.1 | NP_001316015.1 | 220 | 107 | ||
GenBank mRNA | HQ896432.1 | - | 220 | 107 |

More Information
Set Membership: |
![]() ![]() |
