Assay Details
Gene Symbol: CC2D2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001349481.1 | NP_001336410.1 | 11 - 12 | 1276 | 104 | |
XM_017348448.1 | XP_017203937.1 | 2 - 3 | 343 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
