Assay Details
Gene Symbol: CC2D2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001349481.1 | NP_001336410.1 | 25 - 26 | 3114 | 144 | |
XM_017348448.1 | XP_017203937.1 | 16 - 17 | 2181 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() |
