Assay Details
Gene Symbol: APOE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082643.1 | NP_001076112.1 | 36 | 68 | ||
GenBank mRNA | M36603.1 | - | 36 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001082643.1 transcripts.
Set Membership: |
![]() ![]() |
