Assay Details
Gene Symbol: SLF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008261942.2 | XP_008260164.1 | 19 - 20 | 2640 | 73 | |
XM_017344600.1 | XP_017200089.1 | 19 - 20 | 2609 | 73 | ||
XM_017344601.1 | XP_017200090.1 | 20 - 21 | 2686 | 73 | ||
XM_017344602.1 | XP_017200091.1 | 19 - 20 | 2588 | 73 | ||
XM_017344603.1 | XP_017200092.1 | 18 - 19 | 2489 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
