Assay Details
Gene Symbol: FAM155A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002713024.3 | XP_002713070.1 | 2 - 3 | 1520 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM155A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002713024.3 | XP_002713070.1 | 2 - 3 | 1520 | 87 |
Set Membership: |
![]() ![]() ![]() ![]() |