Assay Details
Gene Symbol: KANSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002711192.3 | XP_002711238.1 | 6 - 7 | 974 | 69 | |
XM_008256402.2 | XP_008254624.1 | 6 - 7 | 1228 | 69 | ||
XM_008256404.2 | XP_008254626.1 | 6 - 7 | 1228 | 69 | ||
XM_017341824.1 | XP_017197313.1 | 5 - 6 | 681 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
