Assay Details
Gene Symbol: MYBPHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008264745.2 | XP_008262967.1 | 7 - 8 | 1612 | 58 | |
XM_008264747.2 | XP_008262969.1 | 7 - 8 | 1604 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
