Assay Details
Gene Symbol: JAZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008261564.2 | XP_008259786.1 | 3 - 4 | 390 | 78 | |
XM_017344401.1 | XP_017199890.1 | 3 - 4 | 286 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
