Assay Details
Gene Symbol: ERG28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002719637.3 | XP_002719683.1 | 2 - 3 | 259 | 76 | |
XM_008272027.2 | XP_008270249.1 | 2 - 3 | 243 | 76 | ||
XM_008272028.2 | XP_008270250.1 | 2 - 3 | 242 | 76 | ||
XM_017349416.1 | XP_017204905.1 | 2 - 3 | 225 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
