Assay Details
Gene Symbol: NCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002709690.3 | XP_002709736.1 | 1 - 2 | 165 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002709690.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
