Assay Details
Gene Symbol: NCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002709690.3 | XP_002709736.1 | 3 - 4 | 594 | 87 | |
XM_017339318.1 | XP_017194807.1 | 3 - 4 | 504 | 87 | ||
XM_017339321.1 | XP_017194810.1 | 3 - 4 | 504 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
