Assay Details
Gene Symbol: LOC100347489 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008260488.2 | XP_008258710.1 | 20 - 21 | 2603 | 94 | |
XM_008260489.2 | XP_008258711.1 | 21 - 22 | 3398 | 94 | ||
XM_008260490.2 | XP_008258712.1 | 20 - 21 | 2456 | 94 | ||
XM_008260491.2 | XP_008258713.1 | 21 - 22 | 2751 | 94 | ||
XM_008260492.2 | XP_008258714.1 | 20 - 21 | 2534 | 94 | ||
XM_008260493.2 | XP_008258715.1 | 20 - 21 | 2341 | 94 | ||
XM_008260494.2 | XP_008258716.1 | 20 - 21 | 2346 | 94 | ||
XM_008260495.2 | XP_008258717.1 | 20 - 21 | 2542 | 94 | ||
XM_017343754.1 | XP_017199243.1 | 20 - 21 | 2369 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
