Assay Details
Gene Symbol: SPOUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002723894.3 | XP_002723940.1 | 1 - 2 | 243 | 80 | |
XM_008252766.2 | XP_008250988.1 | 1 - 2 | 376 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
