Assay Details
Gene Symbol: NBAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002710080.3 | XP_002710126.1 | 47 - 48 | 6275 | 57 | |
XM_008254756.2 | XP_008252978.1 | 47 - 48 | 6169 | 57 | ||
XM_017340893.1 | XP_017196382.1 | 47 - 48 | 6962 | 57 | ||
XM_017340894.1 | XP_017196383.1 | 47 - 48 | 6275 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
