Assay Details
Gene Symbol: GADD45A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002715933.3 | XP_002715979.1 | 2 - 3 | 415 | 51 | |
XM_017346192.1 | XP_017201681.1 | 2 - 3 | 453 | 51 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
