Assay Details
Gene Symbol: LOC100344750 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008267611.2 | XP_008265833.1 | 13 - 14 | 1651 | 102 | |
XM_008267612.2 | XP_008265834.1 | 13 - 14 | 1548 | 102 | ||
XM_017347409.1 | XP_017202898.1 | 9 - 10 | 1045 | 102 | ||
XM_017347410.1 | XP_017202899.1 | 13 - 14 | 1653 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
