Assay Details
Gene Symbol: SPART | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008273554.2 | XP_008271776.1 | 746 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008273554.2 transcripts.
Set Membership: |
![]() ![]() |
