Assay Details
Gene Symbol: SPART | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002720586.3 | XP_002720632.1 | 1355 | 108 | ||
XM_008273553.2 | XP_008271775.1 | 1123 | 108 | |||
XM_008273554.2 | XP_008271776.1 | 1755 | 108 |

More Information
Set Membership: |
![]() ![]() |
