Assay Details
Gene Symbol: KLHDC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002715213.3 | XP_002715259.1 | 1 - 2 | 647 | 91 | |
XM_017345739.1 | XP_017201228.1 | 1 - 2 | 647 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
