Assay Details
Gene Symbol: ALG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008261528.2 | XP_008259750.1 | 14 - 15 | 1805 | 56 | |
XM_008261532.2 | XP_008259754.1 | 14 - 15 | 1804 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
