Assay Details
Gene Symbol: PTTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002710352.2 | XP_002710398.1 | 1 - 2 | 162 | 96 | |
XM_008255353.1 | XP_008253575.1 | 1 - 2 | 162 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002710352.2, XM_008255353.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
