Assay Details
Gene Symbol: RNF128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002720143.3 | XP_002720189.1 | 6 - 7 | 1389 | 70 | |
XM_008272843.2 | XP_008271065.1 | 6 - 7 | 1320 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
