Assay Details
Gene Symbol: TENT4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008275053.2 | XP_008273275.1 | 1434 | 81 | ||
XM_008275054.2 | XP_008273276.1 | 1435 | 81 | |||
XM_008275056.2 | XP_008273278.1 | 1305 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
