Assay Details
Gene Symbol: TENT4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008275053.2 | XP_008273275.1 | 279 | 95 | ||
XM_008275054.2 | XP_008273276.1 | 280 | 95 | |||
XM_008275056.2 | XP_008273278.1 | 291 | 95 |

More Information
Set Membership: |
![]() ![]() |
