Assay Details
Gene Symbol: DMAC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002718233.3 | XP_002718279.1 | 2 - 3 | 127 | 90 | |
XM_008269635.2 | XP_008267857.1 | 2 - 3 | 226 | 90 | ||
XM_008269636.2 | XP_008267858.1 | 2 - 3 | 109 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002718233.3, XM_008269635.2, XM_008269636.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
