Assay Details
Gene Symbol: C2CD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008258970.2 | XP_008257192.1 | 10 - 11 | 1157 | 101 | |
XM_017343030.1 | XP_017198519.1 | 10 - 11 | 1157 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
