Assay Details
Gene Symbol: CDKN2AIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002709488.3 | XP_002709534.3 | 2 - 3 | 782 | 103 | |
XM_017338377.1 | XP_017193866.1 | 2 - 3 | 795 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
