Assay Details
Gene Symbol: THEGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008267820.1 | XP_008266042.1 | 5 - 6 | 881 | 88 | |
XM_008267821.1 | XP_008266043.1 | 4 - 5 | 752 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
