Assay Details
Gene Symbol: SENP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008268776.2 | XP_008266998.1 | 1 - 2 | 403 | 118 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008268776.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
