Assay Details
Gene Symbol: C14H3orf38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008267097.2 | XP_008265319.1 | 1 - 2 | 520 | 62 | |
XM_008267098.2 | XP_008265320.1 | 1 - 2 | 523 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
