Assay Details
Gene Symbol: PHYHIPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008270105.2 | XP_008268327.2 | 2 - 3 | 565 | 96 | |
XM_008270106.2 | XP_008268328.1 | 2 - 3 | 323 | 96 | ||
XM_008270107.2 | XP_008268329.1 | 2 - 3 | 685 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
