Assay Details
Gene Symbol: TP53TG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008274760.2 | XP_008272982.1 | 760 | 101 | ||
XM_008274761.2 | XP_008272983.1 | 760 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
