Assay Details
Gene Symbol: CCDC189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008248721.2 | XP_008246943.1 | 1357 | 86 | ||
XM_008248722.2 | XP_008246944.1 | 1325 | 86 | |||
XM_008248723.2 | XP_008246945.1 | 1282 | 86 | |||
XM_008248725.2 | XP_008246947.1 | 1291 | 86 | |||
XM_008248727.2 | XP_008246949.1 | 1240 | 86 | |||
XM_008248729.2 | XP_008246951.1 | 1204 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
