Assay Details
Gene Symbol: RTF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002720873.3 | XP_002720919.1 | 827 | 100 | ||
XM_008274142.2 | XP_008272364.1 | 818 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
