Assay Details
Gene Symbol: ADNP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008252907.2 | XP_008251129.1 | 1876 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008252907.2 transcripts.
Set Membership: |
![]() ![]() |
