Assay Details
Gene Symbol: ADNP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008252907.2 | XP_008251129.1 | 2896 | 99 | ||
XM_017340083.1 | XP_017195572.1 | 141 | 99 | |||
XM_017340084.1 | XP_017195573.1 | 1228 | 99 |

More Information
Set Membership: |
![]() ![]() |
