Assay Details
Gene Symbol: SLC33A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002716312.3 | XP_002716358.1 | 3 - 4 | 1432 | 84 | |
XM_008266376.2 | XP_008264598.1 | 3 - 4 | 1432 | 84 | ||
XM_008266377.2 | XP_008264599.1 | 3 - 4 | 1432 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
