Assay Details
Gene Symbol: KBTBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002708515.3 | XP_002708561.1 | 1 - 2 | 234 | 110 | |
XM_008261949.2 | XP_008260171.1 | 1 - 2 | 282 | 110 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002708515.3, XM_008261949.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
