Assay Details
Gene Symbol: LOC100348238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008262893.2 | XP_008261115.1 | 11 - 12 | 1500 | 115 | |
XM_008262894.2 | XP_008261116.1 | 11 - 12 | 1501 | 115 | ||
XM_008262895.2 | XP_008261117.1 | 11 - 12 | 1726 | 115 | ||
XM_008262899.2 | XP_008261121.1 | 12 - 13 | 1558 | 115 | ||
XM_008262901.2 | XP_008261123.1 | 12 - 13 | 1531 | 115 | ||
XM_017345107.1 | XP_017200596.1 | 11 - 12 | 1500 | 115 | ||
XM_017345108.1 | XP_017200597.1 | 11 - 12 | 1501 | 115 | ||
XM_017345109.1 | XP_017200598.1 | 10 - 11 | 1308 | 115 | ||
XM_017345110.1 | XP_017200599.1 | 11 - 12 | 1504 | 115 | ||
XM_017345111.1 | XP_017200600.1 | 11 - 12 | 1503 | 115 | ||
XM_017345112.1 | XP_017200601.1 | 11 - 12 | 1503 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
