Assay Details
Gene Symbol: LOC100348238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008262893.2 | XP_008261115.1 | 29 - 30 | 4414 | 98 | |
XM_008262894.2 | XP_008261116.1 | 28 - 29 | 4289 | 98 | ||
XM_008262895.2 | XP_008261117.1 | 29 - 30 | 4640 | 98 | ||
XM_008262899.2 | XP_008261121.1 | 30 - 31 | 4472 | 98 | ||
XM_008262901.2 | XP_008261123.1 | 30 - 31 | 4445 | 98 | ||
XM_017345107.1 | XP_017200596.1 | 29 - 30 | 4411 | 98 | ||
XM_017345108.1 | XP_017200597.1 | 29 - 30 | 4394 | 98 | ||
XM_017345109.1 | XP_017200598.1 | 28 - 29 | 4222 | 98 | ||
XM_017345112.1 | XP_017200601.1 | 28 - 29 | 4298 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
