Assay Details
Gene Symbol: MTFR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002710531.3 | XP_002710577.1 | 5 - 6 | 832 | 60 | |
XM_008255583.2 | XP_008253805.1 | 5 - 6 | 834 | 60 | ||
XM_017341350.1 | XP_017196839.1 | 4 - 5 | 695 | 60 | ||
XM_017341351.1 | XP_017196840.1 | 6 - 7 | 934 | 60 | ||
XM_017341352.1 | XP_017196841.1 | 2 - 3 | 267 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
