Assay Details
Gene Symbol: DNAI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008265088.2 | XP_008263310.1 | 3 - 4 | 450 | 141 | |
XM_017346207.1 | XP_017201696.1 | 3 - 4 | 450 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() |
