Assay Details
Gene Symbol: KBTBD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002709099.3 | XP_002709145.1 | 3 - 4 | 837 | 96 | |
XM_002709100.3 | XP_002709146.1 | 3 - 4 | 831 | 96 | ||
XM_008270673.2 | XP_008268895.1 | 3 - 4 | 813 | 96 | ||
XM_017346490.1 | XP_017201979.1 | 3 - 4 | 807 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
