Assay Details
Gene Symbol: BICRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017338829.1 | XP_017194318.1 | 228 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017338829.1 transcripts.
Set Membership: |
![]() ![]() |
