Assay Details
Gene Symbol: ADAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002723773.2 | XP_002723819.1 | 1149 | 66 | ||
XM_008252505.1 | XP_008250727.1 | 1236 | 66 | |||
XM_008252507.1 | XP_008250729.1 | 1101 | 66 |

More Information
Set Membership: |
![]() ![]() |
